Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc27a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077248.1 | NP_001070716.1 | 4 - 5 | 923 | 63 | |
XM_005160200.4 | XP_005160257.1 | 4 - 5 | 960 | 63 | ||
GenBank mRNA | BC076535.1 | - | 3 - 4 | 666 | 63 | |
BC124752.1 | - | 4 - 5 | 923 | 63 | ||
BC164558.1 | - | 3 - 4 | 758 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |