Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkar2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077370.2 | NP_001070838.2 | 7 - 8 | 1254 | 93 | |
GenBank mRNA | BC124725.1 | - | 7 - 8 | 1255 | 93 | |
BC164493.1 | - | 7 - 8 | 894 | 93 | ||
BQ481277.1 | - | 4 - 5 | 370 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |