Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077449.1 | NP_001070917.1 | 43 - 44 | 7424 | 72 | |
XM_009304756.3 | XP_009303031.1 | 44 - 45 | 7925 | 72 | ||
XM_009304757.3 | XP_009303032.1 | 44 - 45 | 7926 | 72 | ||
GenBank mRNA | DQ086492.1 | - | 43 - 44 | 7424 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |