Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nop53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079946.1 | NP_001073415.1 | 11 - 12 | 1344 | 112 | |
GenBank mRNA | BC109456.1 | - | 11 - 12 | 1344 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |