Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ano5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079983.1 | NP_001073452.1 | 15 - 16 | 2177 | 96 | |
XM_005168891.4 | XP_005168948.1 | 16 - 17 | 2314 | 96 | ||
XM_009303225.3 | XP_009301500.1 | 14 - 15 | 1980 | 96 | ||
GenBank mRNA | BC128870.1 | - | 15 - 16 | 2177 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |