Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrxn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079984.1 | NP_001073453.1 | 6 - 7 | 2914 | 58 | |
GenBank mRNA | DQ641430.1 | - | 6 - 7 | 2914 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |