Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03136893_m1
See other NRXN1A GE Assays ›
Gene Symbol:
nrxn1a
Gene Name:
neurexin 1a
Gene Aliases:
-
Chromosome Location:
Chr.12: 114926 - 403053 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03136893_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: nrxn1a
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001080021.2
NP_001073490.1
21 - 22
5079
80
NM_001346923.1
NP_001333852.1
5 - 6
1971
80
NM_001346924.1
NP_001333853.1
4 - 5
1881
80
XM_005156209.4
XP_005156266.1
21 - 22
5073
80
XM_009306641.3
XP_009304916.1
22 - 23
5100
80
XM_009306642.3
XP_009304917.1
22 - 23
5097
80
XM_009306643.3
XP_009304918.1
22 - 23
5099
80
XM_009306644.3
XP_009304919.1
22 - 23
5088
80
XM_009306645.3
XP_009304920.1
21 - 22
5088
80
XM_009306646.3
XP_009304921.1
21 - 22
5085
80
XM_009306648.3
XP_009304923.1
21 - 22
5082
80
XM_009306649.3
XP_009304924.1
21 - 22
5076
80
XM_009306650.3
XP_009304925.1
21 - 22
5073
80
XM_009306651.3
XP_009304926.1
21 - 22
5060
80
XM_009306652.3
XP_009304927.1
21 - 22
5010
80
XM_009306653.3
XP_009304928.1
21 - 22
5010
80
XM_009306654.3
XP_009304929.1
20 - 21
4971
80
XM_009306656.3
XP_009304931.1
20 - 21
4971
80
XM_021480136.1
XP_021335811.1
20 - 21
5070
80
XM_021480137.1
XP_021335812.1
21 - 22
5060
80
XM_021480138.1
XP_021335813.1
19 - 20
5040
80
XM_021480139.1
XP_021335814.1
20 - 21
5034
80
XM_021480140.1
XP_021335815.1
19 - 20
5024
80
XM_021480141.1
XP_021335816.1
19 - 20
4944
80
XM_021480142.1
XP_021335817.1
15 - 16
4860
80
XM_021480143.1
XP_021335818.1
19 - 20
5030
80
XM_021480144.1
XP_021335819.1
17 - 18
4973
80
XM_021480145.1
XP_021335820.1
15 - 16
4860
80
XM_021480146.1
XP_021335821.1
16 - 17
4884
80
GenBank mRNA
AB214179.1
-
6 - 7
1479
80
AB214180.1
-
5 - 6
1389
80
DQ641424.1
-
20 - 21
5028
80
DQ641425.1
-
5 - 6
1971
80
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
cell adhesion
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0