Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080193.1 | NP_001073662.1 | 25 - 26 | 3707 | 63 | |
XM_005168796.4 | XP_005168853.1 | 26 - 27 | 6543 | 63 | ||
GenBank mRNA | BC066564.1 | - | 25 - 26 | 4589 | 63 | |
BC129198.1 | - | 25 - 26 | 3707 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |