Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080193.1 | NP_001073662.1 | 8 - 9 | 901 | 66 | |
XM_005168796.4 | XP_005168853.1 | 8 - 9 | 3602 | 66 | ||
GenBank mRNA | BC066564.1 | - | 8 - 9 | 901 | 66 | |
BC129198.1 | - | 8 - 9 | 901 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |