Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: d2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080597.1 | NP_001074066.1 | 2 - 3 | 410 | 125 | |
XM_005168417.4 | XP_005168474.1 | 2 - 3 | 481 | 125 | ||
GenBank mRNA | BC129361.1 | - | 2 - 3 | 432 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |