Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: d2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080597.1 | NP_001074066.1 | 4 - 5 | 604 | 75 | |
XM_005168417.4 | XP_005168474.1 | 4 - 5 | 675 | 75 | ||
GenBank mRNA | BC129361.1 | - | 4 - 5 | 626 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |