Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083566.1 | NP_001077035.1 | 3 - 4 | 351 | 74 | |
XM_005169631.3 | XP_005169688.1 | 4 - 5 | 477 | 74 | ||
XM_021481055.1 | XP_021336730.1 | 3 - 4 | 334 | 74 | ||
GenBank mRNA | BC115182.1 | - | 3 - 4 | 351 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |