Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083865.1 | NP_001077334.1 | 9 - 10 | 1119 | 84 | |
NM_131049.1 | NP_571124.1 | 11 - 12 | 1663 | 84 | ||
XM_005168429.4 | XP_005168486.1 | 11 - 12 | 1652 | 84 | ||
XM_017359054.2 | XP_017214543.1 | 11 - 12 | 1631 | 84 | ||
XM_021481386.1 | XP_021337061.1 | 10 - 11 | 1593 | 84 | ||
GenBank mRNA | AF073289.1 | - | 11 - 12 | 1663 | 84 | |
BC078280.1 | - | 10 - 11 | 1268 | 84 | ||
BC155168.1 | - | 11 - 12 | 1626 | 84 | ||
CU458994.1 | - | 9 - 10 | 1119 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |