Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098488.1 | NP_001091958.1 | 14 - 15 | 2093 | 68 | |
XM_021468823.1 | XP_021324498.1 | 13 - 14 | 2013 | 68 | ||
XM_021468824.1 | XP_021324499.1 | 14 - 15 | 2145 | 68 | ||
XM_021468825.1 | XP_021324500.1 | 14 - 15 | 2224 | 68 | ||
XM_021468826.1 | XP_021324501.1 | 14 - 15 | 2031 | 68 | ||
XM_021468827.1 | XP_021324502.1 | 13 - 14 | 2013 | 68 | ||
XM_021468828.1 | XP_021324503.1 | 13 - 14 | 2013 | 68 | ||
XM_021468829.1 | XP_021324504.1 | 13 - 14 | 2013 | 68 | ||
XM_021468830.1 | XP_021324505.1 | 13 - 14 | 2013 | 68 | ||
XM_021468831.1 | XP_021324506.1 | 13 - 14 | 2013 | 68 | ||
XM_021468832.1 | XP_021324507.1 | 13 - 14 | 2013 | 68 | ||
XM_021468833.1 | XP_021324508.1 | 13 - 14 | 2013 | 68 | ||
XM_021468834.1 | XP_021324509.1 | 13 - 14 | 2013 | 68 | ||
GenBank mRNA | EF375552.1 | - | 14 - 15 | 2093 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |