Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098488.1 | NP_001091958.1 | 40 - 41 | 5448 | 65 | |
XM_021468823.1 | XP_021324498.1 | 38 - 39 | 5353 | 65 | ||
XM_021468824.1 | XP_021324499.1 | 40 - 41 | 5500 | 65 | ||
XM_021468825.1 | XP_021324500.1 | 40 - 41 | 5579 | 65 | ||
XM_021468826.1 | XP_021324501.1 | 40 - 41 | 5386 | 65 | ||
XM_021468827.1 | XP_021324502.1 | 38 - 39 | 5335 | 65 | ||
XM_021468828.1 | XP_021324503.1 | 38 - 39 | 5320 | 65 | ||
XM_021468829.1 | XP_021324504.1 | 38 - 39 | 5308 | 65 | ||
XM_021468830.1 | XP_021324505.1 | 38 - 39 | 5353 | 65 | ||
XM_021468831.1 | XP_021324506.1 | 39 - 40 | 5368 | 65 | ||
XM_021468832.1 | XP_021324507.1 | 37 - 38 | 5287 | 65 | ||
XM_021468833.1 | XP_021324508.1 | 38 - 39 | 5308 | 65 | ||
XM_021468834.1 | XP_021324509.1 | 37 - 38 | 5293 | 65 | ||
GenBank mRNA | EF375552.1 | - | 40 - 41 | 5448 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |