Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110025.1 | NP_001103495.1 | 3 - 4 | 394 | 66 | |
XM_021478689.1 | XP_021334364.1 | 3 - 4 | 479 | 66 | ||
GenBank mRNA | BC153314.1 | - | 3 - 4 | 394 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |