Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131014.1 | NP_571089.1 | 6 - 7 | 344 | 74 | |
XM_005156049.4 | XP_005156106.1 | 6 - 7 | 420 | 74 | ||
XM_005156050.4 | XP_005156107.1 | 5 - 6 | 377 | 74 | ||
XM_009306481.3 | XP_009304756.1 | 6 - 7 | 420 | 74 | ||
XM_009306483.3 | XP_009304758.1 | 6 - 7 | 420 | 74 | ||
XM_009306485.3 | XP_009304760.1 | 6 - 7 | 420 | 74 | ||
XM_017358425.2 | XP_017213914.1 | 6 - 7 | 420 | 74 | ||
XM_017358426.2 | XP_017213915.1 | 6 - 7 | 420 | 74 | ||
XM_021480020.1 | XP_021335695.1 | 6 - 7 | 420 | 74 | ||
XM_021480021.1 | XP_021335696.1 | 6 - 7 | 420 | 74 | ||
GenBank mRNA | AF102865.1 | - | 6 - 7 | 344 | 74 | |
BC090694.1 | - | 6 - 7 | 393 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |