Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131284.2 | NP_571359.2 | 1 - 2 | 237 | 64 | |
GenBank mRNA | AF052250.1 | - | 1 - 2 | 192 | 64 | |
AY247950.1 | - | 1 - 2 | 73 | 64 | ||
BC056569.1 | - | 1 - 2 | 210 | 64 | ||
BC065668.1 | - | 1 - 2 | 210 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |