Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glra4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131782.2 | NP_571857.1 | 2 - 3 | 710 | 120 | |
XM_005169598.4 | XP_005169655.1 | 2 - 3 | 641 | 120 | ||
GenBank mRNA | AJ404970.1 | - | 2 - 3 | 714 | 120 | |
BC090676.1 | - | 2 - 3 | 365 | 120 | ||
GQ406229.1 | - | 2 - 3 | 198 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |