Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cry5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131788.1 | NP_571863.1 | 2 - 3 | 409 | 105 | |
XM_005155405.3 | XP_005155462.1 | 3 - 4 | 546 | 105 | ||
GenBank mRNA | AB042254.1 | - | 2 - 3 | 409 | 105 | |
BC044204.1 | - | 3 - 4 | 488 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |