Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: selenot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178292.5 | NP_840077.3 | 4 - 5 | 555 | 93 | |
GenBank mRNA | AY216586.1 | - | 4 - 5 | 455 | 93 | |
BC053147.1 | - | 4 - 5 | 502 | 93 | ||
CK870132.1 | - | 4 - 5 | 522 | 93 | ||
CN014436.1 | - | 4 - 5 | 587 | 93 | ||
EH502237.1 | - | 4 - 5 | 546 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |