Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328507.1 | NP_001315436.1 | 3 - 4 | 319 | 81 | |
XM_017353804.1 | XP_017209293.1 | 3 - 4 | 337 | 81 | ||
GenBank mRNA | AY391423.1 | - | 3 - 4 | 248 | 81 | |
AY648844.1 | - | 3 - 4 | 261 | 81 | ||
BC049413.1 | - | 3 - 4 | 258 | 81 | ||
BC071488.1 | - | 3 - 4 | 280 | 81 | ||
CT612850.2 | - | 3 - 4 | 298 | 81 | ||
EH439441.1 | - | 3 - 4 | 327 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |