Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328507.1 | NP_001315436.1 | 6 - 7 | 592 | 102 | |
XM_017353804.1 | XP_017209293.1 | 6 - 7 | 610 | 102 | ||
GenBank mRNA | AY391423.1 | - | 6 - 7 | 521 | 102 | |
AY648844.1 | - | 6 - 7 | 534 | 102 | ||
BC049413.1 | - | 6 - 7 | 531 | 102 | ||
BC071488.1 | - | 6 - 7 | 553 | 102 | ||
CT612850.2 | - | 6 - 7 | 571 | 102 | ||
EH439441.1 | - | 6 - 7 | 600 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |