Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cel.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199607.2 | NP_955901.2 | 3 - 4 | 351 | 91 | |
GenBank mRNA | BC055668.1 | - | 3 - 4 | 351 | 91 | |
BC079529.1 | - | 3 - 4 | 342 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |