Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: castor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199780.2 | NP_956074.2 | 4 - 5 | 828 | 67 | |
GenBank mRNA | BC045430.1 | - | 4 - 5 | 828 | 67 | |
BC057472.1 | - | 4 - 5 | 601 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |