Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199818.1 | NP_956112.1 | 2 - 3 | 529 | 150 | |
GenBank mRNA | BC045850.1 | - | 2 - 3 | 529 | 150 | |
BC075783.1 | - | 2 - 3 | 530 | 150 | ||
BC152679.1 | - | 2 - 3 | 529 | 150 | ||
BC164028.1 | - | 2 - 3 | 233 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |