Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gclm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199845.1 | NP_956139.1 | 6 - 7 | 735 | 82 | |
XM_009303802.3 | XP_009302077.1 | 6 - 7 | 820 | 82 | ||
GenBank mRNA | BC044532.1 | - | 6 - 7 | 735 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |