Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200338.1 | NP_956632.1 | 7 - 8 | 1299 | 62 | |
GenBank mRNA | BC053128.1 | - | 7 - 8 | 1299 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |