Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tle5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200423.1 | NP_956717.1 | 2 - 3 | 249 | 63 | |
XM_005170443.4 | XP_005170500.1 | 2 - 3 | 522 | 63 | ||
GenBank mRNA | BC054644.1 | - | 2 - 3 | 249 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |