Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200737.1 | NP_957031.1 | 3 - 4 | 304 | 67 | |
GenBank mRNA | BC059522.1 | - | 3 - 4 | 304 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |