Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200859.1 | NP_957153.1 | 3 - 4 | 458 | 95 | |
GenBank mRNA | BC062851.1 | - | 3 - 4 | 458 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |