Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201206.2 | NP_957500.2 | 5 - 6 | 855 | 73 | |
GenBank mRNA | BC046017.1 | - | 5 - 6 | 842 | 73 | |
BC066452.1 | - | 5 - 6 | 832 | 73 | ||
EH439430.1 | - | 5 - 6 | 867 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |