Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coro1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201295.1 | NP_958452.1 | 4 - 5 | 573 | 83 | |
GenBank mRNA | BC045340.1 | - | 4 - 5 | 573 | 83 | |
BC165745.1 | - | 3 - 4 | 480 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |