Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201313.1 | NP_958470.1 | 10 - 11 | 1376 | 70 | |
GenBank mRNA | BC047809.1 | - | 10 - 11 | 1376 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |