Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207057.1 | NP_996940.1 | 10 - 11 | 1434 | 137 | |
GenBank mRNA | BC066393.1 | - | 10 - 11 | 1434 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |