Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207057.1 | NP_996940.1 | 12 - 13 | 1630 | 81 | |
XM_005161589.4 | XP_005161646.1 | 12 - 13 | 1630 | 81 | ||
XM_005161590.4 | XP_005161647.1 | 12 - 13 | 1588 | 81 | ||
XM_005161591.4 | XP_005161648.1 | 12 - 13 | 1585 | 81 | ||
GenBank mRNA | BC056523.1 | - | 12 - 13 | 1620 | 81 | |
BC066393.1 | - | 12 - 13 | 1630 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |