Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ilf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212599.1 | NP_997764.1 | 3 - 4 | 328 | 95 | |
XM_005163739.4 | XP_005163796.1 | 3 - 4 | 400 | 95 | ||
XM_005163740.4 | XP_005163797.1 | 3 - 4 | 400 | 95 | ||
XM_005163741.3 | XP_005163798.1 | 3 - 4 | 400 | 95 | ||
XM_005163742.4 | XP_005163799.1 | 3 - 4 | 400 | 95 | ||
XM_009299225.3 | XP_009297500.1 | 3 - 4 | 400 | 95 | ||
GenBank mRNA | BC047175.1 | - | 3 - 4 | 357 | 95 | |
BC067172.1 | - | 3 - 4 | 328 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |