Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214714.1 | NP_999879.1 | 5 - 6 | 829 | 64 | |
XM_009302506.2 | XP_009300781.1 | 5 - 6 | 901 | 64 | ||
GenBank mRNA | BC046876.1 | - | 5 - 6 | 845 | 64 | |
BC068350.1 | - | 5 - 6 | 829 | 64 | ||
BC165318.1 | - | 5 - 6 | 790 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |