Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002122.2 | NP_001002122.2 | 2 - 3 | 424 | 146 | |
GenBank mRNA | BC049428.1 | - | 2 - 3 | 396 | 146 | |
BC071463.1 | - | 2 - 3 | 373 | 146 | ||
EH504663.1 | - | 2 - 3 | 437 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |