Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002122.2 | NP_001002122.2 | 9 - 10 | 1298 | 145 | |
GenBank mRNA | BC071463.1 | - | 9 - 10 | 1247 | 145 | |
EH459652.1 | - | 1 - 2 | 115 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |