Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002124.2 | NP_001002124.1 | 5 - 6 | 564 | 108 | |
GenBank mRNA | BC071453.1 | - | 5 - 6 | 566 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |