Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002124.2 | NP_001002124.1 | 6 - 7 | 610 | 94 | |
GenBank mRNA | BC071453.1 | - | 6 - 7 | 612 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |