Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002357.1 | NP_001002357.1 | 7 - 8 | 493 | 98 | |
XM_021466675.1 | XP_021322350.1 | 5 - 6 | 1100 | 98 | ||
GenBank mRNA | BC075898.1 | - | 7 - 8 | 493 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |