Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkx6.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002475.1 | NP_001002475.1 | 2 - 3 | 1033 | 87 | |
GenBank mRNA | AY437556.1 | - | 2 - 3 | 1025 | 87 | |
BC076337.1 | - | 2 - 3 | 1033 | 87 | ||
BC164096.1 | - | 2 - 3 | 723 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |