Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crhbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003459.1 | NP_001003459.1 | 4 - 5 | 590 | 95 | |
GenBank mRNA | BC076472.1 | - | 4 - 5 | 590 | 95 | |
BC164122.1 | - | 4 - 5 | 586 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |