Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpdb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003742.1 | NP_001003742.1 | 8 - 9 | 566 | 72 | |
GenBank mRNA | BC077167.1 | - | 8 - 9 | 566 | 72 | |
BC165880.1 | - | 7 - 8 | 557 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |