Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005293.1 | NP_001005293.1 | 9 - 10 | 1593 | 67 | |
GenBank mRNA | AB354736.1 | - | 9 - 10 | 1554 | 67 | |
AY648760.1 | - | 9 - 10 | 1593 | 67 | ||
BC116516.1 | - | 9 - 10 | 1575 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |