Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006094.1 | NP_001006094.1 | 10 - 11 | 882 | 89 | |
XM_005157422.4 | XP_005157479.1 | 10 - 11 | 934 | 89 | ||
GenBank mRNA | BC083224.1 | - | 10 - 11 | 882 | 89 | |
BC163990.1 | - | 9 - 10 | 749 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |