Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spopla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013447.1 | NP_001013465.1 | 5 - 6 | 618 | 114 | |
GenBank mRNA | BC090815.1 | - | 5 - 6 | 618 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |