Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017755.1 | NP_001017755.1 | 2 - 3 | 131 | 66 | |
GenBank mRNA | BC093359.1 | - | 2 - 3 | 131 | 66 | |
BC165450.1 | - | 2 - 3 | 153 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |